Learn More
Cayman Chemical Human Fibroblast Growth Factor 1c/α-Klotho (FGFR1c/α-Klotho) for Endocrine FGF Signaling Reporter Assay System, 384 Well
SDP
Catalog No. 502942749
Encompass_Preferred
An all-inclusive cell-based luciferase reporter assay kit targeting the Human Fibroblast Growth Factor Receptor 1c and α-Klotho (FGFR/α-Klotho); utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the Human Fibroblast Growth Factor Receptor 1c and α-Klotho; provides two optimized media for use during cell culture and in diluting the user’s test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate
Catalog No. 50-294-2749 Supplier Cayman Chemical Supplier No. 42742384 WELL
An all-inclusive cell-based luciferase reporter assay kit targeting the Human Fibroblast Growth Factor Receptor 1c and α-Klotho (FGFR/α-Klotho); utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the Human Fibroblast Growth Factor Receptor 1c and α-Klotho; provides two optimized media for use during cell culture and in diluting the user’s test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate