Learn More
Cayman Chemical Human G Protein-Coupled Bile Acid Receptor 1 Reporter Assay System, 96 Well
SDP
Catalog No. 502945946
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to TGR5 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Lithocholic Acid (LCA), Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-5946 Supplier Cayman Chemical Supplier No. 4101796 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to TGR5 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Lithocholic Acid (LCA), Luciferase Detection Reagents, and a cell culture-ready assay plate.