Learn More
Cayman Chemical Humn GlucocortIcoId Receptr 96
SDP
Catalog No. 502969800
Encompass_Preferred
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00201-32, IB00201, IB00202]
Catalog No. 50-296-9800 Supplier Cayman Chemical Supplier No. 1573396 WELL
Add to Cart
Add to Cart
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00201-32, IB00201, IB00202]