Learn More
Cayman Chemical Humn GlucocortIcoId RECPTR
SDP
Catalog No. 502971909
Encompass_Preferred
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00201-32, IB00201, IB00202]
Catalog No. 50-297-1909 Supplier Cayman Chemical Supplier No. 157333X32 WELL
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00201-32, IB00201, IB00202]