Learn More
Cayman Chemical Human Growth Hormone Receptor, isoform 1 Reporter Assay System, 384 Well
SDP
Catalog No. 502947728
Encompass_Preferred
Human Growth Hormone Receptor, isoform 1 Reporter Cells are prepared using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. IB14001, IB14001-32, and IB14002]
Catalog No. 50-294-7728 Supplier Cayman Chemical Supplier No. 33508384 WELL
Human Growth Hormone Receptor, isoform 1 Reporter Cells are prepared using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. IB14001, IB14001-32, and IB14002]