Learn More
Cayman Chemical Human Growth Hormone Receptor, isoform 1 Reporter Assay System, 96 Well
SDP
Catalog No. 502948541
Encompass_Preferred
Human Growth Hormone Receptor, isoform 1 Reporter Cells are prepared using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. IB14001, IB14001-32, and IB14002]
Catalog No. 50-294-8541 Supplier Cayman Chemical Supplier No. 3350896 WELL
Human Growth Hormone Receptor, isoform 1 Reporter Cells are prepared using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. IB14001, IB14001-32, and IB14002]