Learn More
Cayman Chemical Humn InsulIn ReceptrIsofo 384
SDP
Catalog No. 502944900
Encompass_Preferred
A cell-based luciferase reporter assay for human insulin receptor, isoform B (INSRb)
Catalog No. 50-294-4900 Supplier Cayman Chemical Supplier No. 41021384 WELL
A cell-based luciferase reporter assay for human insulin receptor, isoform B (INSRb)