Learn More
Cayman Chemical Human Insulin-like Growth Factor-1 Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502944826
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for IGF-1R signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator Insulin, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-4826 Supplier Cayman Chemical Supplier No. 4102296 WELL
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for IGF-1R signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator Insulin, Luciferase Detection Reagents, and a cell culture-ready assay plate.