Learn More
Cayman Chemical Humn LIver X ReceptrAlpha 96
SDP
Catalog No. 502968616
Encompass_Preferred
LXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00311-32, IB00311, IB00312]
Catalog No. 50-296-8616 Supplier Cayman Chemical Supplier No. 1573596 WELL
LXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00311-32, IB00311, IB00312]