Learn More
Cayman Chemical Humn LIver X ReceptrBeta 96Wel
SDP
Catalog No. 502971912
Encompass_Preferred
LXRβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00301-32, IB00301, IB00302]
Catalog No. 50-297-1912 Supplier Cayman Chemical Supplier No. 1573496 WELL
LXRβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00301-32, IB00301, IB00302]