Learn More
Cayman Chemical Human Liver Receptor Homolog-1 Reporter Assay System, 96 Well
SDP
Catalog No. 502984753
Encompass_Preferred
LRH-1 Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB11001-32, IB11001, IB11002]
Catalog No. 50-298-4753 Supplier Cayman Chemical Supplier No. 1910996 WELL
LRH-1 Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB11001-32, IB11001, IB11002]