Learn More
Cayman Chemical Human Liver X Receptor, Alpha Reporter Assay System, 3x32 Well
SDP
Catalog No. 502971913
Encompass_Preferred
LXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00311-32, IB00311, IB00312]
Catalog No. 50-297-1913 Supplier Cayman Chemical Supplier No. 157353X32 WELL
LXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00311-32, IB00311, IB00312]