Learn More
Cayman Chemical Humn Macrophage CoLNyStImu 96
SDP
Catalog No. 502943008
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for M-CSFR signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator M-CSF as a positive control, Luciferase Detection Reagents, and a cell culture-ready assay plate
Catalog No. 50-294-3008 Supplier Cayman Chemical Supplier No. 4006896 WELL
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for M-CSFR signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator M-CSF as a positive control, Luciferase Detection Reagents, and a cell culture-ready assay plate