Learn More
Cayman Chemical Humn MInralocortIcoId Recep
SDP
Supplier:  Cayman Chemical 157403X32 WELL
Encompass_Preferred
MR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00501-32, IB00501, IB00502]
Catalog No. 50-296-8879