Learn More
Cayman Chemical Humn MInralocortIcoId Recep 96
SDP
Catalog No. 502972586
Encompass_Preferred
MR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00501-32, IB00501, IB00502]
Catalog No. 50-297-2586 Supplier Cayman Chemical Supplier No. 1574096 WELL
MR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00501-32, IB00501, IB00502]