Learn More
Cayman Chemical Human Androgen Receptor Reporter Assay System, 384 Well
SDP
Catalog No. 502969829
Encompass_Preferred
AR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB03001-32, IB03001, IB03002]
Catalog No. 50-296-9829 Supplier Cayman Chemical Supplier No. 15756384 WELL
AR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB03001-32, IB03001, IB03002]