Learn More
Cayman Chemical Humn ndrogen RECPTR Repor
SDP
Catalog No. 502972683
Encompass_Preferred
AR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB03001-32, IB03001, IB03002]
Catalog No. 50-297-2683 Supplier Cayman Chemical Supplier No. 157563X32 WELL
AR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB03001-32, IB03001, IB03002]