Learn More
Cayman Chemical Human Nuclear Factor (erythroi-derived 2)-like 2 Reporter Assay System, 3x32 Well
SDP
Catalog No. 502968219
Encompass_Preferred
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay wells. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments prior to assay setup. Reporter Cells are single use reagents. They can not be refrozen or maintained in extended culture. [INDIGO Catalog Nos. IB10001-32, IB10001, IB10002]
Catalog No. 50-296-8219 Supplier Cayman Chemical Supplier No. 177343X32 WELL
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay wells. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments prior to assay setup. Reporter Cells are single use reagents. They can not be refrozen or maintained in extended culture. [INDIGO Catalog Nos. IB10001-32, IB10001, IB10002]