Learn More
Cayman Chemical Human Oxytocin Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502942534
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to OXTR Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Oxytocin, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-2534 Supplier Cayman Chemical Supplier No. 4089096 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to OXTR Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Oxytocin, Luciferase Detection Reagents, and a cell culture-ready assay plate.