Learn More
Cayman Chemical Humn OxytocIn RECPTR Repor 384
SDP
Catalog No. 502943164
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to OXTR Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Oxytocin, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-3164 Supplier Cayman Chemical Supplier No. 40890384 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to OXTR Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator Oxytocin, Luciferase Detection Reagents, and a cell culture-ready assay plate.