Learn More
Cayman Chemical Humn PGlycoproteIn MDR1 D 96
SDP
Supplier:  Cayman Chemical 2833996 WELL
Encompass_Preferred
The materials provided in the assay kit for MDR1 / Human P-Glycoprotein Drug Interaction are formatted to allow for two independent 48-well assay setups. EAch aliquot of HCT-Pgp Cells is provided as a single-use assay reagent and provides sufficient volume to dispense 48 assay wells of HCT-Pgp cells into one 96-well plate. If desired, the two aliquots of HCT-Pgp cells may be thawed, combined to generate a single cell suspension, and then dispensed into all wells of an assay plate. The kit contains two aliquots of HCT-Pgp cells and two black, sterile, collagen-coated 96-well assay plates. In addition to HCT-Pgp cells and assay plates, this kit includes Cell Recovery Medium (CRM-p) used to perform a rapid thaw of the -80°C HCT-Pgp cells, Compound Screening Medium supplemented with daunorubicin (CSM+DR) used to prepare drug treatment media, Wash Buffer, two aliquots of Lysis Reagent, and the potent reference inhibitor Tariquidar.
Catalog No. 50-294-0919