Learn More
Cayman Chemical Human Pregnane X Receptor Reporter Assay System, 3x32 Well
SDP
Catalog No. 502966837
Encompass_Preferred
PXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. IB07001-32, IB07001, IB07002]
Catalog No. 50-296-6837 Supplier Cayman Chemical Supplier No. 175283X32 WELL
PXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. IB07001-32, IB07001, IB07002]