Learn More
Cayman Chemical Human Progesterone Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502972571
Encompass_Preferred
PGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB05001-32, IB05001, IB05002]
Catalog No. 50-297-2571 Supplier Cayman Chemical Supplier No. 1575996 WELL
PGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB05001-32, IB05001, IB05002]