Learn More
Cayman Chemical Human Peroxisome Proliferator-Activated Receptor Beta/Delta Reporter Assay System, 3x32 Well
SDP
Catalog No. 502972517
Encompass_Preferred
PPAR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00121-32, IB00121, IB00122]
Catalog No. 50-297-2517 Supplier Cayman Chemical Supplier No. 157313X32 WELL
PPAR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00121-32, IB00121, IB00122]