Learn More
Cayman Chemical Human Peroxisome Proliferator-Activated Receptor Alpha Reporter Assay System, 3x32 Well
SDP
Catalog No. 502972536
Encompass_Preferred
PPARα Reporter Assay Systems utilize non-human mammalian cells engineered to express human PPARA, commonly referred to as PPARα.[INDIGO Catalog Nos. IB00111-32, IB00111, IB00112]
Catalog No. 50-297-2536 Supplier Cayman Chemical Supplier No. 157303X32 WELL
PPARα Reporter Assay Systems utilize non-human mammalian cells engineered to express human PPARA, commonly referred to as PPARα.[INDIGO Catalog Nos. IB00111-32, IB00111, IB00112]