Learn More
Cayman Chemical Human Peroxisome Proliferator-Activated Receptor Gamma Reporter Assay System, 3x32 Well
SDP
Catalog No. 502972591
Encompass_Preferred
PPARγ Reporter Assay Systems utilize non-human mammalian cells engineered to express human PPARG, commonly referred to as PPARγ [INDIGO Catalog Nos. IB00101-32, IB00101, IB00102]
Catalog No. 50-297-2591 Supplier Cayman Chemical Supplier No. 157293X32 WELL
PPARγ Reporter Assay Systems utilize non-human mammalian cells engineered to express human PPARG, commonly referred to as PPARγ [INDIGO Catalog Nos. IB00101-32, IB00101, IB00102]