Learn More
Cayman Chemical Human Peroxisome Proliferator-Activated Receptor Panel, 3x32 Well
SDP
Catalog No. 502972516
Encompass_Preferred
This PANEL contains sufficient materials to perform 32 PPARα assays, 32 PPARγ assays, and 32 PPARδ assays, all in a single 96-well plate format. [INDIGO Catalog No. IB00131-32P]
Catalog No. 50-297-2516 Supplier Cayman Chemical Supplier No. 157323X32 WELL
This PANEL contains sufficient materials to perform 32 PPARα assays, 32 PPARγ assays, and 32 PPARδ assays, all in a single 96-well plate format. [INDIGO Catalog No. IB00131-32P]