Learn More
Cayman Chemical Human Retinoic Acid Receptor Gamma Reporter Assay System, 3x32 Well
SDP
Catalog No. 502972609
Encompass_Preferred
RARγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB02001-32, IB02001, IB02002]
Catalog No. 50-297-2609 Supplier Cayman Chemical Supplier No. 157523X32 WELL
RARγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB02001-32, IB02001, IB02002]