Learn More
Cayman Chemical Humn RetInoIc ACD Receptr 3x32
SDP
Catalog No. 502972201
Encompass_Preferred
RARβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB02101-32, IB02101, IB02102]
Catalog No. 50-297-2201 Supplier Cayman Chemical Supplier No. 157533X32 WELL
RARβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB02101-32, IB02101, IB02102]