Learn More
Cayman Chemical Human Retinoic Acid Receptor Panel, 96 Well
SDP
Catalog No. 502972577
Encompass_Preferred
RAR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog No. IB02301-48P]
Catalog No. 50-297-2577 Supplier Cayman Chemical Supplier No. 1575596 WELL
RAR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog No. IB02301-48P]