Learn More
Cayman Chemical Human Retinoid X Receptor Beta Reporter Assay System, 96 Well
SDP
Catalog No. 502968884
Encompass_Preferred
RXRβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00811-32, IB00811, IB00812]
Catalog No. 50-296-8884 Supplier Cayman Chemical Supplier No. 1574496 WELL
RXRβ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00811-32, IB00811, IB00812]