Learn More
Cayman Chemical Humn RetInoId X RECPTR Gam
SDP
Catalog No. 502971923
Encompass_Preferred
RXRγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00821-32, IB00821, IB00822]
Catalog No. 50-297-1923 Supplier Cayman Chemical Supplier No. 157453X32 WELL
Add to Cart
Add to Cart
RXRγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00821-32, IB00821, IB00822]