Learn More
Cayman Chemical Humn RPP30-DNA Control asy 1ea
SDP
Catalog No. 502943985
Encompass_Preferred
A mix of primers and probe targeting human RPP30DNA Intron I (20X concentrated) as a PCR positive control assay for human samples. The primers amplify the intron region of the gene, this assay will only detect amplification from genomic DNA and will not detect spliced mRNA. Organism Type: Human Enzyme Master Mixes (recommended): Empirical qPCR mastermix, highQU ORA qPCR enzyme mastermix, Standard qPCR enzyme mastermix (Item No. 40706) Panels: Control Colors: HEX (RPP30) Instruments: Any instrument
Catalog No. 50-294-3985 Supplier Cayman Chemical Supplier No. 406821 EA
A mix of primers and probe targeting human RPP30DNA Intron I (20X concentrated) as a PCR positive control assay for human samples. The primers amplify the intron region of the gene, this assay will only detect amplification from genomic DNA and will not detect spliced mRNA. Organism Type: Human Enzyme Master Mixes (recommended): Empirical qPCR mastermix, highQU ORA qPCR enzyme mastermix, Standard qPCR enzyme mastermix (Item No. 40706) Panels: Control Colors: HEX (RPP30) Instruments: Any instrument