Learn More
Cayman Chemical Human Erythropoietin Receptor Reporter Assay System, 384 Well
SDP
Catalog No. 502947664
Encompass_Preferred
Reporter cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB17001, IB17001-32, IB17002]
Catalog No. 50-294-7664 Supplier Cayman Chemical Supplier No. 34027384 WELL
Reporter cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB17001, IB17001-32, IB17002]