Learn More
Cayman Chemical Human Estrogen Receptor Alpha Reporter Assay System, 384 Well
SDP
Catalog No. 502968615
Encompass_Preferred
ER Alpha Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00401-32, IB00401, IB00402]
Catalog No. 50-296-8615 Supplier Cayman Chemical Supplier No. 15737384 WELL
ER Alpha Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00401-32, IB00401, IB00402]