Learn More
Cayman Chemical Human Estrogen Receptor Beta Reporter Assay System, 96 Well
SDP
Catalog No. 502971911
Encompass_Preferred
ER Beta Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00411-32, IB00411, IB00412]
Catalog No. 50-297-1911 Supplier Cayman Chemical Supplier No. 1573896 WELL
ER Beta Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB00411-32, IB00411, IB00412]