Learn More
Cayman Chemical Human Tropomysin Receptor Kinase C Reporter Assay System, 96 Well
SDP
Catalog No. 502944749
Encompass_Preferred
A cell-based luciferase reporter assay for human TrkC
Catalog No. 50-294-4749 Supplier Cayman Chemical Supplier No. 4102096 WELL
A cell-based luciferase reporter assay for human TrkC