Learn More
Cayman Chemical Human Tropomysin Receptor Kinase A Reporter Assay System, 96 Well
SDP
Catalog No. 502944989
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for Human TrkA. In addition to the Reporter Cells, this kit includes an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator NGF-β, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-4989 Supplier Cayman Chemical Supplier No. 4101896 WELL
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for Human TrkA. In addition to the Reporter Cells, this kit includes an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator NGF-β, Luciferase Detection Reagents, and a cell culture-ready assay plate.