Learn More
Cayman Chemical Human Tropomysin Receptor Kinase B Reporter Assay System, 384 Well
SDP
Catalog No. 502944665
Encompass_Preferred
INDIGO’s Reporter Cells are transiently transfected and prepared as frozen stocks using a proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations or cell titer adjustments prior to assay setup. This assay kit provides the convenience of an allinclusive cell-based reporter assay system for Human TrkB. In addition to the Reporter Cells, this kit includes an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator NT-4, Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-4665 Supplier Cayman Chemical Supplier No. 41019384 WELL
INDIGO’s Reporter Cells are transiently transfected and prepared as frozen stocks using a proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations or cell titer adjustments prior to assay setup. This assay kit provides the convenience of an allinclusive cell-based reporter assay system for Human TrkB. In addition to the Reporter Cells, this kit includes an optimized culture medium for use in reviving the cryopreserved cells, a culture medium for use in preparing test sample treatments, the physiological activator NT-4, Luciferase Detection Reagents, and a cell culture-ready assay plate.