Learn More
Cayman Chemical Humn TropomysIn RECPTR KIn 384
SDP
Catalog No. 502942827
Encompass_Preferred
A cell-based luciferase reporter assay for human TrkC
Catalog No. 50-294-2827 Supplier Cayman Chemical Supplier No. 41020384 WELL
A cell-based luciferase reporter assay for human TrkC