Learn More
Cayman Chemical Human Vascular Endothelial Growth Factor Receptor 2 Reporter Assay System, 96 Well
SDP
Catalog No. 502947673
Encompass_Preferred
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to VEGFR2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator human VEGF-A (165 aa isoform, non-glycosylated), Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-7673 Supplier Cayman Chemical Supplier No. 3350996 WELL
INDIGO's assay kits provide the convenience of an all-inclusive cell-based assay system. In addition to VEGFR2 Reporter Cells, provided are two optimized media for use in recovering the cryopreserved cells and for diluting test samples, the reference activator human VEGF-A (165 aa isoform, non-glycosylated), Luciferase Detection Reagents, and a cell culture-ready assay plate.