Learn More
Cayman Chemical Humn VItamIn D RECPTR Repo
SDP
Catalog No. 502969835
Encompass_Preferred
VDR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00701-32, IB00701, IB00702]
Catalog No. 50-296-9835 Supplier Cayman Chemical Supplier No. 157423X32 WELL
VDR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. IB00701-32, IB00701, IB00702]