Learn More
Bellco Glass, Inc. Hungate Tube Complete, 16X125mm
SDP
Catalog No. 501945338
Encompass_Preferred
The hungate anaerobic culture tube was developed for growing and maintaining anaerobic bacteria and culture conditions. method of hungate utilizes syringe methods for anaerobic. MADE IN THE USA. •Hungate Culture Tubes are designed for maintaining anaerobic culture conditions•Dimensions: 16mm OD x 125mm OAL•Material: Type 1 Class A Borosilicate•Case Quantity: 250•Screw caps included, catalog #2047-16000•Septum stoppers included, catalog #2047-11600Technique: Exclude oxygen by flushing the tube with the desired gas. Add 4.5mL od pre-reduced anaerobic agar medium into tube. Seal the tube with the butyl rubber stopper and screw cap and autoclave. Innoculate with a syringe. Incubate in water bath.
Catalog No. 50-194-5338 Supplier Bellco Glass, Inc. Supplier No. 204716125
Add to Cart
Add to Cart
The hungate anaerobic culture tube was developed for growing and maintaining anaerobic bacteria and culture conditions. method of hungate utilizes syringe methods for anaerobic. MADE IN THE USA. •Hungate Culture Tubes are designed for maintaining anaerobic culture conditions•Dimensions: 16mm OD x 125mm OAL•Material: Type 1 Class A Borosilicate•Case Quantity: 250•Screw caps included, catalog #2047-16000•Septum stoppers included, catalog #2047-11600Technique: Exclude oxygen by flushing the tube with the desired gas. Add 4.5mL od pre-reduced anaerobic agar medium into tube. Seal the tube with the butyl rubber stopper and screw cap and autoclave. Innoculate with a syringe. Incubate in water bath.