Learn More
Chemglass Life Sciences Complete Hungate Anaerobic Tubes, 16 x 125mm, Supplied Complete with Butyl Stoppper and Cap with 9mm Opening
SDP
Catalog No. 501215187
Encompass_Preferred
Complete Hungate Anaerobic Tubes, 16 x 125mm, Supplied Complete with Butyl Stopper and Cap with 9mm Opening. 16 x 125mm screw thread style tube designed to maintain anaerobic culture conditions. Flange style stoppers are made from butyl rubber. Complete item consists of a disposable glass tube, butyl stopper and screw cap having a 9mm opening.
Catalog No. 50-121-5187 Supplier Chemglass Life Sciences Supplier No. CLS420801
Complete Hungate Anaerobic Tubes, 16 x 125mm, Supplied Complete with Butyl Stopper and Cap with 9mm Opening. 16 x 125mm screw thread style tube designed to maintain anaerobic culture conditions. Flange style stoppers are made from butyl rubber. Complete item consists of a disposable glass tube, butyl stopper and screw cap having a 9mm opening.