Learn More
Bellco Glass, Inc. Hungate Tube Only,16x125mm
SDP
Catalog No. 501945335
Encompass_Preferred
The Hungate Anaerobic Culture Tube was developed for growing and maintaining anaerobic bacteria and culture conditions. method of hungate utilizes syringe methods for anaerobic. MADE IN THE USA.Technique:Exclude oxygen by flushing the tube with the desired gasAdd 4.5mL of pre-reduced anaerobic agar medium into tubeSeal the tube with the butyl rubber stopper and screw cap and autoclaveInoculate with a syringeIncubate in a water bathSpecifications:Neck: 15-415Volume:16.5mlDimension: 16x125mmCase Quantity: 250
Catalog No. 50-194-5335 Supplier Bellco Glass, Inc. Supplier No. 204700125
Add to Cart
Add to Cart
The Hungate Anaerobic Culture Tube was developed for growing and maintaining anaerobic bacteria and culture conditions. method of hungate utilizes syringe methods for anaerobic. MADE IN THE USA.Technique:Exclude oxygen by flushing the tube with the desired gasAdd 4.5mL of pre-reduced anaerobic agar medium into tubeSeal the tube with the butyl rubber stopper and screw cap and autoclaveInoculate with a syringeIncubate in a water bathSpecifications:Neck: 15-415Volume:16.5mlDimension: 16x125mmCase Quantity: 250