Learn More
Chemglass Life Sciences Tube Only, Hungate Anaerobic, 16 x 125mm, White SBS cell Shrink Wrapped
SDP
Catalog No. 501215188
Encompass_Preferred
Tube Only, Hungate Anaerobic, 16 x 125mm, White SBS cell Shrink Wrapped. 16 x 125mm screw thread style tube designed to maintain anaerobic culture conditions. Flange style stoppers are made from butyl rubber. Complete item consists of a disposable glass tube, butyl stopper and screw cap having a 9mm opening.
Catalog No. 50-121-5188 Supplier Chemglass Life Sciences Supplier No. CLS420810
Tube Only, Hungate Anaerobic, 16 x 125mm, White SBS cell Shrink Wrapped. 16 x 125mm screw thread style tube designed to maintain anaerobic culture conditions. Flange style stoppers are made from butyl rubber. Complete item consists of a disposable glass tube, butyl stopper and screw cap having a 9mm opening.