Learn More
Enzo Life Sciences Huntingtin polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW0595A0100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 2-17 of human huntingtin. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-1546