Learn More
MSC Huot CNC Tool Cart, 36 Tool Capacity, 50 Taper Size
Catalog No. 5000515666
Encompass
This CNC tool cart is designed to efficiently store and transport tooling within industrial environments. Constructed from durable steel, it features a 36-tool capacity and accommodates 50 taper size tools, including BT, CAT V Flange, and NMTB types. The cart's robust design supports a load capacity of 1,000 lbs, ensuring secure handling of heavy loads.
  • Holds 36 tools.
  • Accommodates BT, CAT V Flange, and NMTB 50 taper tools.
  • Supports a load capacity of 1,000 lbs.
  • Equipped with two fixed and two swivel casters with locks.
  • Constructed from steel.
  • Features 5 in polyurethane casters.
  • Measures 35-1/4 in depth, 35 in height, and 18 in width.
  • Available in red and black colors.
Catalog No. 50-005-15666 Supplier MSC Supplier No. 85153047
This CNC tool cart is designed to efficiently store and transport tooling within industrial environments. Constructed from durable steel, it features a 36-tool capacity and accommodates 50 taper size tools, including BT, CAT V Flange, and NMTB types. The cart's robust design supports a load capacity of 1,000 lbs, ensuring secure handling of heavy loads.
  • Holds 36 tools.
  • Accommodates BT, CAT V Flange, and NMTB 50 taper tools.
  • Supports a load capacity of 1,000 lbs.
  • Equipped with two fixed and two swivel casters with locks.
  • Constructed from steel.
  • Features 5 in polyurethane casters.
  • Measures 35-1/4 in depth, 35 in height, and 18 in width.
  • Available in red and black colors.