Learn More
MSC Huot Drill Bit Case, For Jobber Bits, Straight Shank, 1/16 to 1/2", Plastic
Catalog No. 5000530364
Encompass
The Huot 1/16 to 1/2" Drill Bit Case is designed for organizing and protecting jobber length, straight shank drill bits. Made from durable plastic, it features a water-resistant and break-resistant construction, making it suitable for various work environments. It can store up to 29 drill bits.
  • Water-resistant construction
  • Break-resistant design
  • Features a quarter turn cover
  • Includes a clip-on cup
  • Compatible with jobber length drill bits
  • Compatible with straight shank drill bits
  • Stores up to 29 drill bits
  • Designed for fractional inch sizes with 1/64" step increments
Catalog No. 50-005-30364 Supplier MSC Supplier No. 65164873
The Huot 1/16 to 1/2" Drill Bit Case is designed for organizing and protecting jobber length, straight shank drill bits. Made from durable plastic, it features a water-resistant and break-resistant construction, making it suitable for various work environments. It can store up to 29 drill bits.
  • Water-resistant construction
  • Break-resistant design
  • Features a quarter turn cover
  • Includes a clip-on cup
  • Compatible with jobber length drill bits
  • Compatible with straight shank drill bits
  • Stores up to 29 drill bits
  • Designed for fractional inch sizes with 1/64" step increments