Learn More
MSC Huot Drill Bit Index, Steel, Lockable, 9/16 to 1 in
Catalog No. 5000529615
Encompass
This drill bit index is designed for Silver & Deming drill bits, offering compatibility with reduced shank drill bits. It features 1/16 in step increments and dimensions of 8-1/2 in wide x 2 in high x 6-1/4 in deep. Constructed from rugged, heavy gauge steel, it is corrosion-resistant and has an epoxy baked finish. It includes a secure cover and precisely sized holes to keep drill bits organized and protected for a range of 9/16 to 1 inch drill bits.
  • Lockable for added security.
  • Compatible with reduced shank drill bits.
  • Designed for 1/2 in shank drill bits.
  • Accommodates 8 popular sizes from 9/16 to 1 in.
  • Constructed from rugged, heavy gauge steel.
  • Corrosion-resistant with an epoxy baked finish.
  • Includes a secure cover and precisely sized holes.
Catalog No. 50-005-29615 Supplier MSC Supplier No. 01058338
This drill bit index is designed for Silver & Deming drill bits, offering compatibility with reduced shank drill bits. It features 1/16 in step increments and dimensions of 8-1/2 in wide x 2 in high x 6-1/4 in deep. Constructed from rugged, heavy gauge steel, it is corrosion-resistant and has an epoxy baked finish. It includes a secure cover and precisely sized holes to keep drill bits organized and protected for a range of 9/16 to 1 inch drill bits.
  • Lockable for added security.
  • Compatible with reduced shank drill bits.
  • Designed for 1/2 in shank drill bits.
  • Accommodates 8 popular sizes from 9/16 to 1 in.
  • Constructed from rugged, heavy gauge steel.
  • Corrosion-resistant with an epoxy baked finish.
  • Includes a secure cover and precisely sized holes.